Product Details

SNP ID
rs7305594
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:121717127 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCATGCTGGTGAGTTGAGTGCTAGT[C/T]TGAGACCAGTGTGCTCCTGGTCTCC
Phenotype
MIM: 616551
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMEM120B PubMed Links

Gene Details

Gene
TMEM120B
Gene Name
transmembrane protein 120B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080825.2 Intron NP_001074294.2
XM_005253843.4 Intron XP_005253900.1
XM_017018853.1 Intron XP_016874342.1
XM_017018854.1 Intron XP_016874343.1

View Full Product Details