Product Details

SNP ID
rs7179979
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:78287938 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCTTGAAAATGGATGCTCTAATTT[C/T]TTCCTGTCCCCAGGGGCCAGCTACA
Phenotype
MIM: 609540
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNAJA4 PubMed Links

Gene Details

Gene
DNAJA4
Gene Name
DnaJ heat shock protein family (Hsp40) member A4
There are no transcripts associated with this gene.

Gene
WDR61
Gene Name
WD repeat domain 61
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303247.1 Intron NP_001290176.1
NM_001303248.1 Intron NP_001290177.1
NM_025234.2 Intron NP_079510.1
XM_011522094.1 Intron XP_011520396.1
XM_017022637.1 Intron XP_016878126.1

View Full Product Details