Product Details

SNP ID
rs6713743
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:69916952 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCATAGTTTCTGGTACATGCAAAT[A/T]GTCCATCAATGTTGCTGCCCGTTTC
Phenotype
MIM: 600021
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
MXD1 PubMed Links

Gene Details

Gene
MXD1
Gene Name
MAX dimerization protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202513.1 Intron NP_001189442.1
NM_001202514.1 Intron NP_001189443.1
NM_002357.3 Intron NP_002348.1

View Full Product Details