Product Details

SNP ID
rs6661941
Assay Type
Functionally tested
NCBI dbSNP Submissions
28
Location
Chr.1:27536544 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCTGCTGCTGGGATGAATGCAAGC[G/T]CTCCACAGTCAGGCGGTAAACAGGC
Phenotype
MIM: 615790
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
AHDC1 PubMed Links

Gene Details

Gene
AHDC1
Gene Name
AT-hook DNA binding motif containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001029882.3 Intron NP_001025053.1
XM_005245848.3 Intron XP_005245905.1
XM_005245849.3 Intron XP_005245906.1
XM_005245850.3 Intron XP_005245907.1
XM_005245851.3 Intron XP_005245908.1
XM_005245852.3 Intron XP_005245909.1
XM_011541255.1 Intron XP_011539557.1
XM_011541256.2 Intron XP_011539558.1
XM_011541257.2 Intron XP_011539559.1

View Full Product Details