Product Details
- SNP ID
-
hCV29332224
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:142245006 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGCTCACAATAGGGAGAGGAAGAAA[A/G]GTGCAGGCATACGTGGTGTCATCCT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MOXD2P
PubMed Links
Gene Details
- Gene
- MOXD2P
- Gene Name
- monooxygenase, DBH-like 2, pseudogene
There are no transcripts associated with this gene.
- Gene
- PRSS58
- Gene Name
- protease, serine 58
There are no transcripts associated with this gene.
View Full Product Details