Product Details

SNP ID
rs6919110
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:17600388 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAAACGGAGCAGTGACAGGTATCCC[A/G]GAGGGTGCTGCTGAGGCGACGATGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM8A1 PubMed Links

Gene Details

Gene
FAM8A1
Gene Name
family with sequence similarity 8 member A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016255.2 102 UTR 5 NP_057339.1

View Full Product Details