Product Details

SNP ID
rs6940818
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:17601596 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGTTAGGAAGTTTTGATTGGGTCA[A/G]AGCTTCACTTCATTTGATCAATATC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM8A1 PubMed Links

Gene Details

Gene
FAM8A1
Gene Name
family with sequence similarity 8 member A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016255.2 Intron NP_057339.1

View Full Product Details