Product Details

SNP ID
rs7253091
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:37476312 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGGAATATCCACAGCATGACAAAA[C/T]TCTACTTTTTTTTTTCGTATTTGCA
Phenotype
MIM: 613904
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF569 PubMed Links

Gene Details

Gene
ZNF569
Gene Name
zinc finger protein 569
There are no transcripts associated with this gene.

Gene
ZNF570
Gene Name
zinc finger protein 570
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300993.2 Intron NP_001287922.1
NM_001321991.1 Intron NP_001308920.1
NM_001321992.1 Intron NP_001308921.1
NM_001321993.1 Intron NP_001308922.1
NM_001321994.1 Intron NP_001308923.1
NM_144694.3 Intron NP_653295.1
XM_011526544.2 Intron XP_011524846.1
XM_011526545.2 Intron XP_011524847.1

View Full Product Details