Product Details

SNP ID
rs7038960
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:37439078 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTTTGGTATGTCTAAGAAAGCTA[C/T]ATAAAACATAAAATGAACTCATGTT
Phenotype
MIM: 604296 MIM: 616590
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GRHPR PubMed Links

Gene Details

Gene
GRHPR
Gene Name
glyoxylate reductase/hydroxypyruvate reductase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012203.1 3674 Intron NP_036335.1
Gene
ZBTB5
Gene Name
zinc finger and BTB domain containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014872.2 3674 UTR 3 NP_055687.1
XM_005251634.1 3674 UTR 3 XP_005251691.1

View Full Product Details