Product Details

SNP ID
rs7866144
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:101473487 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTAAATGGAGGAAGATCTAGAAGTG[A/G]GTGGGAACAAAAGAGCAGAACTGAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMEM246 PubMed Links

Gene Details

Gene
TMEM246
Gene Name
transmembrane protein 246
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303107.1 3956 UTR 3 NP_001290036.1
NM_001303108.1 3956 UTR 3 NP_001290037.1
NM_032342.2 3956 UTR 3 NP_115718.1
XM_017015206.1 3956 UTR 3 XP_016870695.1
Gene
TMEM246-AS1
Gene Name
TMEM246 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details