Product Details

SNP ID
rs7242527
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:54908213 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTTGTTAAAGATACCCACAATATC[A/G]ATCACATGTTCGTCTTTCTTTTGCG
Phenotype
MIM: 616909
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC68 PubMed Links

Gene Details

Gene
CCDC68
Gene Name
coiled-coil domain containing 68
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143829.1 Intron NP_001137301.1
NM_025214.2 Intron NP_079490.1
XM_006722552.3 Intron XP_006722615.1
XM_011526198.2 Intron XP_011524500.1
XM_011526199.2 Intron XP_011524501.1
XM_011526200.2 Intron XP_011524502.1
XM_011526201.2 Intron XP_011524503.1
XM_011526202.1 Intron XP_011524504.1
XM_011526203.2 Intron XP_011524505.1
XM_011526204.2 Intron XP_011524506.1
XM_017026011.1 Intron XP_016881500.1

View Full Product Details