Product Details

SNP ID
rs6573651
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:24265924 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTAGGCAGGGACTGCAAATGCCT[A/G]CCCACAGGGTCAGCCAGGAAAGAAT
Phenotype
MIM: 601905 MIM: 190195
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RABGGTA PubMed Links

Gene Details

Gene
RABGGTA
Gene Name
Rab geranylgeranyltransferase alpha subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004581.5 Intron NP_004572.3
NM_182836.2 Intron NP_878256.1
Gene
TGM1
Gene Name
transglutaminase 1
There are no transcripts associated with this gene.

View Full Product Details