Product Details

SNP ID
rs7941147
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:34481922 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATTATGCCCAGCACATAGCAATTG[C/T]TTTCTACACACACATTTGTGAAATT
Phenotype
MIM: 115500 MIM: 605169
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CAT PubMed Links

Gene Details

Gene
CAT
Gene Name
catalase
There are no transcripts associated with this gene.

Gene
ELF5
Gene Name
E74 like ETS transcription factor 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243080.1 Intron NP_001230009.1
NM_001243081.1 Intron NP_001230010.1
NM_001422.3 Intron NP_001413.1
NM_198381.1 Intron NP_938195.1
XM_017017308.1 Intron XP_016872797.1
XM_017017309.1 Intron XP_016872798.1

View Full Product Details