Product Details

SNP ID
rs8109417
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:7608212 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCTCCAGGACCTGGCCAGTGGGGC[C/T]GCGCTGGCCGCCACCATCCACTGCT
Phenotype
MIM: 612685
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CAMSAP3 PubMed Links

Gene Details

Gene
CAMSAP3
Gene Name
calmodulin regulated spectrin associated protein family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080429.2 863 Silent Mutation GCC,GCT A263A NP_001073898.1
NM_020902.1 863 Silent Mutation GCC,GCT A236A NP_065953.1
XM_005272491.3 863 Silent Mutation GCC,GCT A252A XP_005272548.1
XM_011528154.2 863 Silent Mutation GCC,GCT A263A XP_011526456.1
XM_017027036.1 863 Silent Mutation GCC,GCT A263A XP_016882525.1
Gene
MIR6792
Gene Name
microRNA 6792
There are no transcripts associated with this gene.

View Full Product Details