Product Details

SNP ID
rs6603107
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:7359234 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAAGGGGAGTGACCATCAGAGGGT[C/T]ATATCAAAAGGACACACCAACTCCA
Phenotype
MIM: 616432
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARHGEF18 PubMed Links

Gene Details

Gene
ARHGEF18
Gene Name
Rho/Rac guanine nucleotide exchange factor 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130955.1 Intron NP_001124427.1
NM_015318.3 Intron NP_056133.2
XM_005272464.4 Intron XP_005272521.1
XM_006722705.3 Intron XP_006722768.1
XM_006722706.3 Intron XP_006722769.1
XM_006722708.2 Intron XP_006722771.1
XM_011527835.2 Intron XP_011526137.1
XM_011527836.2 Intron XP_011526138.1
XM_011527837.2 Intron XP_011526139.1
XM_011527838.2 Intron XP_011526140.1
XM_011527839.2 Intron XP_011526141.1
XM_011527840.1 Intron XP_011526142.1
XM_011527841.2 Intron XP_011526143.1

View Full Product Details