Product Details

SNP ID
rs6438176
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:113964527 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATTGCTGGACTGTCATGATAGAG[C/T]TATTACCTTTTGGAATGTCCTTCAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC191 PubMed Links

Gene Details

Gene
CCDC191
Gene Name
coiled-coil domain containing 191
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020817.1 3528 UTR 3 NP_065868.1
XM_005247681.3 3528 UTR 3 XP_005247738.1
XM_006713715.1 3528 UTR 3 XP_006713778.1
XM_011513051.1 3528 UTR 3 XP_011511353.1
XM_011513052.2 3528 UTR 3 XP_011511354.1
XM_011513053.1 3528 UTR 3 XP_011511355.1
XM_011513054.2 3528 UTR 3 XP_011511356.1
XM_017006956.1 3528 UTR 3 XP_016862445.1
XM_017006957.1 3528 UTR 3 XP_016862446.1
XM_017006958.1 3528 UTR 3 XP_016862447.1
XM_017006959.1 3528 UTR 3 XP_016862448.1
Gene
ZDHHC23
Gene Name
zinc finger DHHC-type containing 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320466.1 3528 Intron NP_001307395.1
NM_001320467.1 3528 Intron NP_001307396.1
NM_001320468.1 3528 Intron NP_001307397.1
NM_173570.4 3528 Intron NP_775841.2
XM_005247269.1 3528 Intron XP_005247326.1
XM_005247270.3 3528 Intron XP_005247327.1
XM_006713562.3 3528 Intron XP_006713625.1
XM_011512618.2 3528 Intron XP_011510920.1
XM_011512619.1 3528 Intron XP_011510921.1
XM_017006084.1 3528 Intron XP_016861573.1
XM_017006085.1 3528 Intron XP_016861574.1
XM_017006086.1 3528 Intron XP_016861575.1
XM_017006087.1 3528 Intron XP_016861576.1

View Full Product Details