Product Details

SNP ID
rs6574233
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:75432163 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAGGGCCTTTTGCTGCTCGCCTTC[A/C]CTCTCAGTCTTGGGGCCTTCCCCAT
Phenotype
MIM: 608657
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
JDP2 PubMed Links

Gene Details

Gene
JDP2
Gene Name
Jun dimerization protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135047.1 431 Intron NP_001128519.1
NM_001135048.1 431 Intron NP_001128520.1
NM_001135049.1 431 UTR 5 NP_001128521.1
NM_130469.3 431 Intron NP_569736.1
XM_005267332.4 431 Intron XP_005267389.1
XM_017020971.1 431 Intron XP_016876460.1
XM_017020972.1 431 UTR 5 XP_016876461.1
XM_017020973.1 431 Intron XP_016876462.1
XM_017020974.1 431 Intron XP_016876463.1
XM_017020975.1 431 Intron XP_016876464.1
XM_017020976.1 431 Intron XP_016876465.1
Gene
LOC101928377
Gene Name
uncharacterized LOC101928377
There are no transcripts associated with this gene.

View Full Product Details