Product Details
- SNP ID
-
rs8060206
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:761199 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGTAAGTAACAACCTTTGAGCTCTT[C/T]CTGTTGTGGTGTGGATGGAATCTGC
- Phenotype
-
MIM: 601051
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MIR662
PubMed Links
Gene Details
- Gene
- MIR662
- Gene Name
- microRNA 662
There are no transcripts associated with this gene.
- Gene
- MSLN
- Gene Name
- mesothelin
View Full Product Details