Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001033855.2 | 2586 | Intron | NP_001029027.1 | ||
NM_001033857.2 | 2586 | Intron | NP_001029029.1 | ||
NM_001033858.2 | 2586 | Intron | NP_001029030.1 | ||
NM_001289076.1 | 2586 | Intron | NP_001276005.1 | ||
NM_001289077.1 | 2586 | Intron | NP_001276006.1 | ||
NM_001289078.1 | 2586 | Intron | NP_001276007.1 | ||
NM_001289079.1 | 2586 | Intron | NP_001276008.1 | ||
NM_022487.3 | 2586 | Intron | NP_071932.2 | ||
XM_006717491.3 | 2586 | Intron | XP_006717554.1 | ||
XM_011519616.1 | 2586 | Intron | XP_011517918.1 | ||
XM_011519617.1 | 2586 | Intron | XP_011517919.1 | ||
XM_011519619.1 | 2586 | Intron | XP_011517921.1 | ||
XM_011519620.2 | 2586 | Intron | XP_011517922.1 | ||
XM_011519621.1 | 2586 | Intron | XP_011517923.1 | ||
XM_017016556.1 | 2586 | UTR 3 | XP_016872045.1 | ||
XM_017016557.1 | 2586 | Intron | XP_016872046.1 | ||
XM_017016558.1 | 2586 | Intron | XP_016872047.1 | ||
XM_017016559.1 | 2586 | UTR 3 | XP_016872048.1 | ||
XM_017016560.1 | 2586 | UTR 3 | XP_016872049.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001193424.1 | 2586 | Intron | NP_001180353.1 | ||
NM_001193425.1 | 2586 | Intron | NP_001180354.1 | ||
NM_001193426.1 | 2586 | Intron | NP_001180355.1 | ||
NM_001193427.1 | 2586 | Intron | NP_001180356.1 | ||
NM_024670.3 | 2586 | Intron | NP_078946.1 | ||
XM_006717503.3 | 2586 | Intron | XP_006717566.1 | ||
XM_011519662.2 | 2586 | Intron | XP_011517964.1 | ||
XM_017016637.1 | 2586 | Intron | XP_016872126.1 |