Product Details

SNP ID
rs6965923
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:26659076 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGCTTATTCCACAATTATGTTCGA[C/T]GTGATATCATAATTCTGAAAACTGG
Phenotype
MIM: 605215
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SKAP2 PubMed Links

Gene Details

Gene
SKAP2
Gene Name
src kinase associated phosphoprotein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303468.1 Intron NP_001290397.1
NM_003930.4 Intron NP_003921.2
XM_017012771.1 Intron XP_016868260.1

View Full Product Details