Product Details
- SNP ID
-
rs8081095
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:47735122 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GACCTGGGATGCTGGGTCCCAGGTC[A/C]GAGAGGTGTGGATACCAAACGTGGG
- Phenotype
-
MIM: 604895
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TBX21
PubMed Links
Gene Details
- Gene
- TBX21
- Gene Name
- T-box 21
View Full Product Details