Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001142407.2 | Intron | NP_001135879.1 | |||
NM_001142408.2 | Intron | NP_001135880.1 | |||
NM_001142417.2 | Intron | NP_001135889.1 | |||
NM_001142435.2 | Intron | NP_001135907.1 | |||
NM_001261822.1 | Intron | NP_001248751.1 | |||
NM_003296.3 | Intron | NP_003287.1 | |||
XM_005249349.1 | Intron | XP_005249406.1 | |||
XM_005249350.1 | Intron | XP_005249407.1 | |||
XM_005249351.3 | Intron | XP_005249408.1 | |||
XM_005249352.3 | Intron | XP_005249409.1 | |||
XM_005249353.3 | Intron | XP_005249410.1 | |||
XM_005249356.1 | Intron | XP_005249413.1 | |||
XM_011514841.1 | Intron | XP_011513143.1 | |||
XM_011514842.1 | Intron | XP_011513144.1 | |||
XM_011514843.2 | Intron | XP_011513145.1 | |||
XM_011514844.1 | Intron | XP_011513146.1 |