Product Details

SNP ID
rs6557400
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:155101259 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGGTGTGCCTCATTTTCTTCTAGT[A/C]TTGCCTTCATCTGGGACATTTTTGC
Phenotype
MIM: 604709
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TIAM2 PubMed Links

Gene Details

Gene
TIAM2
Gene Name
T-cell lymphoma invasion and metastasis 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010927.2 Intron NP_001010927.1
NM_012454.3 Intron NP_036586.2

View Full Product Details