Product Details

SNP ID
rs7761517
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:124977194 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACATTAACACTTTCTGACTAGTTGC[A/C]AGATTAACATCAGGGAAAGTACAAA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
RNF217 PubMed Links

Gene Details

Gene
RNF217
Gene Name
ring finger protein 217
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286398.2 Intron NP_001273327.1
NM_152553.4 Intron NP_689766.1
XM_011535494.2 Intron XP_011533796.1
XM_011535495.2 Intron XP_011533797.1
XM_011535496.2 Intron XP_011533798.1
XM_011535499.2 Intron XP_011533801.1
XM_011535500.2 Intron XP_011533802.1
XM_017010317.1 Intron XP_016865806.1

View Full Product Details