Product Details

SNP ID
rs7757936
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:112053905 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCACTCTGTACGTGAGGGTGAAGC[G/T]GGAGAAGGAAGCAGCCTAGAGAGGA
Phenotype
MIM: 603400
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
WISP3 PubMed Links

Gene Details

Gene
WISP3
Gene Name
WNT1 inducible signaling pathway protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003880.3 Intron NP_003871.1
NM_198239.1 Intron NP_937882.1
XM_011536220.1 Intron XP_011534522.1
XM_011536222.2 Intron XP_011534524.2
XM_011536223.2 Intron XP_011534525.1

View Full Product Details