Product Details

SNP ID
rs6927348
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:116636808 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTATCATATTTGTTTTTAATCCTA[C/G]AATTTAAAAAATTGTAGTTATTGCT
Phenotype
MIM: 612647
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RSPH4A PubMed Links

Gene Details

Gene
RSPH4A
Gene Name
radial spoke head 4 homolog A
There are no transcripts associated with this gene.

Gene
ZUFSP
Gene Name
zinc finger with UFM1 specific peptidase domain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145062.2 Intron NP_659499.2
XM_011535565.2 Intron XP_011533867.1
XM_011535567.1 Intron XP_011533869.1
XM_017010393.1 Intron XP_016865882.1
XM_017010394.1 Intron XP_016865883.1
XM_017010395.1 Intron XP_016865884.1
XM_017010396.1 Intron XP_016865885.1

View Full Product Details