Product Details

SNP ID
rs7362232
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:21512064 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAAAAGGGAATGCCCGCCTGGAA[A/G]GTGGCGGCTGCCAGGTCCTGGGCTT
Phenotype
MIM: 604612
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NKX2-2 PubMed Links

Gene Details

Gene
NKX2-2
Gene Name
NK2 homeobox 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002509.3 695 Silent Mutation ACC,ACT T227T NP_002500.1
XM_006723566.3 695 Silent Mutation ACC,ACT T156T XP_006723629.1

View Full Product Details