Product Details

SNP ID
rs6529426
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:130408908 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCTATTAGTTTCTTTTTGCATTTC[A/G]AATAGCTTTTTTAATTAAAATTTTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RBMX2 PubMed Links

Gene Details

Gene
RBMX2
Gene Name
RNA binding motif protein, X-linked 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016024.2 Intron NP_057108.2

View Full Product Details