Product Details

SNP ID
rs10400836
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:22801261 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATGGACATTTCCAGTTCCTTACC[G/A]GTGCACATGACAGAGCTCATTATCC
Phenotype
MIM: 608145
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
NIPA1 PubMed Links

Gene Details

Gene
NIPA1
Gene Name
non imprinted in Prader-Willi/Angelman syndrome 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142275.1 Intron NP_001135747.1
NM_144599.4 Intron NP_653200.2

View Full Product Details