Product Details

SNP ID
rs9260861
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:29977817 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACCATTATTGGGACATACTTTGCA[C/T]ACAATCAGTGTATCTATTTGAAATG
Phenotype
MIM: 615797
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HCG9 PubMed Links

Gene Details

Gene
HCG9
Gene Name
HLA complex group 9 (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details