Product Details

SNP ID
rs10456182
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:55175951 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGGAGATGGAGGAGGCTAGGAGTT[A/G]CTCTTGATTTTCATTTATAAAAGAA
Phenotype
MIM: 602393
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HCRTR2 PubMed Links

Gene Details

Gene
HCRTR2
Gene Name
hypocretin receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001526.3 Intron NP_001517.2
XM_017010798.1 Intron XP_016866287.1

View Full Product Details