Product Details
- SNP ID
-
rs9934930
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:4852037 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCATTCCCTGATTGATTGTCATTTG[A/G]TTGTCATTTTGGTTGCTTCCATCAT
- Phenotype
-
MIM: 610660
MIM: 609771
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GLYR1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs190482396,rs77213183] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GLYR1
- Gene Name
- glyoxylate reductase 1 homolog
There are no transcripts associated with this gene.
- Gene
- UBN1
- Gene Name
- ubinuclein 1
View Full Product Details