Product Details

SNP ID
rs6689338
Assay Type
Functionally tested
NCBI dbSNP Submissions
20
Location
Chr.1:150283248 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTGGGTACTCCAGGAGCTGTTCTA[A/T]AGCCCCTGCTTCTGGACCTATGGAT
Phenotype
MIM: 607629 MIM: 615782
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
APH1A PubMed Links

Gene Details

Gene
APH1A
Gene Name
aph-1 homolog A, gamma-secretase subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077628.2 58 Intron NP_001071096.1
NM_001243771.1 58 Intron NP_001230700.1
NM_001243772.1 58 Intron NP_001230701.1
NM_016022.3 58 Intron NP_057106.2
XM_017001417.1 58 Intron XP_016856906.1
Gene
C1orf54
Gene Name
chromosome 1 open reading frame 54
There are no transcripts associated with this gene.

Gene
CIART
Gene Name
circadian associated repressor of transcription
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300838.1 58 UTR 5 NP_001287767.1
NM_001300839.1 58 Intron NP_001287768.1
NM_001300840.1 58 Intron NP_001287769.1
NM_001300841.1 58 Intron NP_001287770.1
NM_144697.3 58 UTR 5 NP_653298.1
XM_017000378.1 58 UTR 5 XP_016855867.1

View Full Product Details