Product Details

SNP ID
rs6096114
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:50815881 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCTGAGCACGCCTCCCTGGAGCA[A/G]GACGGTGTGGCCTGGTAGGGAAAGC
Phenotype
MIM: 607471
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BCAS4 PubMed Links

Gene Details

Gene
BCAS4
Gene Name
breast carcinoma amplified sequence 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010974.2 Intron NP_001010974.1
NM_017843.4 Intron NP_060313.3
NM_198799.3 Intron NP_942094.2

View Full Product Details