Product Details

SNP ID
rs10040598
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:148986157 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGCCTATTTTACTGCCCGTGTGAA[A/C]TTGGGTATGTTACATAACTGCCTTA
Phenotype
MIM: 608206
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SH3TC2 PubMed Links

Gene Details

Gene
SH3TC2
Gene Name
SH3 domain and tetratricopeptide repeats 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024577.3 22573 UTR 3 NP_078853.2

View Full Product Details