Product Details

SNP ID
rs10199582
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:227987169 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTTAAAAATGCATATTTCTAGGCT[C/T]CACTACCAGAAAGTGATCAGGTCCT
Phenotype
MIM: 611646
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SPHKAP PubMed Links

Gene Details

Gene
SPHKAP
Gene Name
SPHK1 interactor, AKAP domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142644.1 Intron NP_001136116.1
NM_030623.3 Intron NP_085126.2
XM_005246870.4 Intron XP_005246927.1
XM_006712777.3 Intron XP_006712840.1
XM_006712778.3 Intron XP_006712841.1
XM_011511925.2 Intron XP_011510227.1

View Full Product Details