Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001032394.2 | Intron | NP_001027566.1 | |||
NM_001032395.2 | Intron | NP_001027567.1 | |||
NM_020455.5 | Intron | NP_065188.4 | |||
NM_198569.2 | Intron | NP_940971.1 | |||
XM_005267061.3 | Intron | XP_005267118.1 | |||
XM_006715516.2 | Intron | XP_006715579.1 | |||
XM_006715517.2 | Intron | XP_006715580.1 | |||
XM_006715518.2 | Intron | XP_006715581.1 | |||
XM_011535964.1 | Intron | XP_011534266.1 | |||
XM_017011085.1 | Intron | XP_016866574.1 |