Product Details

SNP ID
rs10510752
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:47019963 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATACCTGATTCTTGGTATTAGAATC[C/T]GTTTAGAGGCCCTGACTCAGGTACA
Phenotype
MIM: 612778
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NRADDP PubMed Links

Gene Details

Gene
NRADDP
Gene Name
neurotrophin receptor associated death domain, pseudogene
There are no transcripts associated with this gene.

Gene
SETD2
Gene Name
SET domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014159.6 Intron NP_054878.5
XM_011533631.2 Intron XP_011531933.1
XM_011533632.2 Intron XP_011531934.1
XM_011533633.2 Intron XP_011531935.1
XM_011533634.1 Intron XP_011531936.1
XM_017006270.1 Intron XP_016861759.1

View Full Product Details