Product Details

SNP ID
rs9960367
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:63952945 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTATTGGAAATCAATAGTTTTTCT[C/T]CTTACAAATTTCATGTTTTAAGCAA
Phenotype
MIM: 612086
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HMSD PubMed Links

Gene Details

Gene
HMSD
Gene Name
histocompatibility minor serpin domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001123366.1 Intron NP_001116838.1
XM_011525930.2 Intron XP_011524232.1
XM_017025710.1 Intron XP_016881199.1

View Full Product Details