Product Details

SNP ID
rs10045497
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:75340659 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAAAATCTGAAACGCTTCTGGTCC[A/C]AAGCATTTTGGATAAAGGATACTCA
Phenotype
MIM: 142910
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HMGCR PubMed Links

Gene Details

Gene
HMGCR
Gene Name
3-hydroxy-3-methylglutaryl-CoA reductase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000859.2 Intron NP_000850.1
NM_001130996.1 Intron NP_001124468.1
XM_011543357.1 Intron XP_011541659.1
XM_011543358.1 Intron XP_011541660.1
XM_011543359.1 Intron XP_011541661.1

View Full Product Details