Product Details

SNP ID
rs10407339
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:14614176 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCTGTACTGTCCGCAGCCTGCTGA[A/G]TGGGTGGGTGTCGTGTTGTAAACCC
Phenotype
MIM: 606101 MIM: 616838
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADGRE3 PubMed Links

Gene Details

Gene
ADGRE3
Gene Name
adhesion G protein-coupled receptor E3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289158.1 Intron NP_001276087.1
NM_001289159.1 Intron NP_001276088.1
NM_032571.4 Intron NP_115960.2
XM_011528374.2 Intron XP_011526676.1
XM_017027383.1 Intron XP_016882872.1
Gene
CLEC17A
Gene Name
C-type lectin domain family 17 member A
There are no transcripts associated with this gene.

Gene
SNORA104
Gene Name
small nucleolar RNA, H/ACA box 104
There are no transcripts associated with this gene.

View Full Product Details