Product Details

SNP ID
rs8181769
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:22632168 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTGAGAAAATAATAAGTGCATGAA[A/G]GTCTTTGTACAGGAAACCGTATGAA
Phenotype
MIM: 609858
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ETNK1 PubMed Links

Gene Details

Gene
ETNK1
Gene Name
ethanolamine kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039481.1 Intron NP_001034570.1
NM_018638.4 Intron NP_061108.2
XM_005253420.4 Intron XP_005253477.1
XM_017019580.1 Intron XP_016875069.1
XM_017019581.1 Intron XP_016875070.1
Gene
LOC105369691
Gene Name
uncharacterized LOC105369691
There are no transcripts associated with this gene.

View Full Product Details