Product Details
- SNP ID
-
rs150284621
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:4862702 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTTTTTTTTCTTTCGGCCCTCAGG[G/T]CGGCTGAGCCCCCCAGCCTGCACCC
- Phenotype
-
MIM: 142983
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MSX1
PubMed Links
Gene Details
- Gene
- MSX1
- Gene Name
- msh homeobox 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002448.3 |
706 |
Silent Mutation |
AGG,AGT |
R157S |
NP_002439.2 |
View Full Product Details