Product Details

SNP ID
rs7432995
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:151738426 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GACCTGTCTCTCCAGCTGCCTTTAC[A/T]TTTTTCCTTTGTTTTAACCTTGGAG
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
AADACL2 PubMed Links

Gene Details

Gene
AADACL2
Gene Name
arylacetamide deacetylase like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207365.3 Intron NP_997248.2
XM_011512753.1 Intron XP_011511055.1

View Full Product Details