Product Details
- SNP ID
-
rs9367875
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:9775825 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATTTTACCTGAGAGTGTAGTTCATC[A/G]TCTTCTTCTAAGAGATTGGGATCAT
- Phenotype
-
MIM: 614287
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
OFCC1
PubMed Links
Gene Details
- Gene
- OFCC1
- Gene Name
- orofacial cleft 1 candidate 1
There are no transcripts associated with this gene.
View Full Product Details