Product Details

SNP ID
rs9890026
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:38942676 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAATATATACTTCAATACTCATTT[G/T]TACAAAATCTTTTGTCCACATTTCA
Phenotype
MIM: 609498
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FBXO47 PubMed Links

Gene Details

Gene
FBXO47
Gene Name
F-box protein 47
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008777.2 Intron NP_001008777.2
XM_011524865.2 Intron XP_011523167.1
XM_011524866.2 Intron XP_011523168.1
XM_011524867.2 Intron XP_011523169.1

View Full Product Details