Product Details
- SNP ID
-
rs9907137
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:78232549 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCAGGGGTTGTTTCTTGATCACGCT[C/T]CCCGGGGTTTGGGCCTGGCAGCCCC
- Phenotype
-
MIM: 603352
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
BIRC5
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs11868709] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- BIRC5
- Gene Name
- baculoviral IAP repeat containing 5
There are no transcripts associated with this gene.
- Gene
- TMEM235
- Gene Name
- transmembrane protein 235
View Full Product Details