Product Details
- SNP ID
-
rs9473700
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:49946798 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGTTGCTTCAATTGCTATCTTGCT[C/T]GAATATTTTTTATAAATCTCTTTTA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DEFB133
PubMed Links
Gene Details
- Gene
- DEFB133
- Gene Name
- defensin beta 133
View Full Product Details