Product Details

SNP ID
rs9514828
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:108269025 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTCCTCAACTAGTATCATATTGAG[C/T]GGGGACTTAAAGCCTCAGAAGTCTG
Phenotype
MIM: 603969
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TNFSF13B PubMed Links

Gene Details

Gene
TNFSF13B
Gene Name
tumor necrosis factor superfamily member 13b
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145645.2 Intron NP_001139117.1
NM_006573.4 Intron NP_006564.1

View Full Product Details