Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001204118.1 | 96 | Intron | NP_001191047.1 | ||
NM_207390.3 | 96 | Intron | NP_997273.3 | ||
XM_017026785.1 | 96 | Intron | XP_016882274.1 | ||
XM_017026786.1 | 96 | Intron | XP_016882275.1 | ||
XM_017026787.1 | 96 | Intron | XP_016882276.1 | ||
XM_017026788.1 | 96 | UTR 5 | XP_016882277.1 | ||
XM_017026789.1 | 96 | Intron | XP_016882278.1 | ||
XM_017026790.1 | 96 | Intron | XP_016882279.1 | ||
XM_017026791.1 | 96 | Intron | XP_016882280.1 | ||
XM_017026792.1 | 96 | Intron | XP_016882281.1 | ||
XM_017026793.1 | 96 | Intron | XP_016882282.1 | ||
XM_017026794.1 | 96 | Intron | XP_016882283.1 |